missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAK3 Polyclonal Antibody

Descripción
Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7, NIH-3T3, JC, mouse brain, rat brain. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (99%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PAK proteins are critical effectors that link Rho GTPases to cytoskeleton reorganization and nuclear signaling. PAK proteins, a family of serine/threonine p21-activating kinases, serve as targets for the small GTP binding proteins Cdc42 and RAC and have been implicated in a wide range of biological activities. The alpha, beta and gammaPAK isoforms complex specifically with Rac1 and Cdc42 in their active GTP-bound state, inhibiting their intrinsic GTPase activity leading to their autophosphorylation. There are eight sites of autophosphorylation on gammaPAK, including Ser 19, Ser 141 and Thr 402, and phosphorylation of Ser 141 and Thr 402 is correlated with gammaPAK activation. Once phosphorylated and their affinity for Rac/Cdc42 reduced, the PAK isoforms disassociate from the complex to seek downstream substrates.
Especificaciones
Especificaciones
| Antígeno | PAK3 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.74 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| génica | PAK3 |
| N.º de referencia del gen | O75914, Q61036, Q62829 |
| Alias de gen | 2.7.11.1; adriamycin resistance-associated; ARA; beta-PAK; bPAK; CDC42/RAC effector kinase PAK-B; CDKN1A; hPAK3; MRX30; MRX47; oligophrenin-3; OPHN3; p21 (CDKN1A)-activated kinase 3; p21 (RAC1) activated kinase 3; p21 protein (Cdc42/Rac)-activated kinase 3; p21/Cdc42/Rac1-activated kinase 3 (yeast Ste20-related); p21-activated kinase 3; p65-PAK; PAK beta; Pak3; Pak-3; PAK3beta; Pak65alpha; Pak65beta; Pakb; Serine/threonine-protein kinase PAK 3; Stk4 |
| Símbolos de los genes | PAK3 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?