missing translation for 'onlineSavingsMsg'
Learn More

PeproTech PAI1 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Código de producto. 17885113
Change view
Click to view available options
Cantidad:
2 x 500 μg
25 μg
50 μg
500 μg
Tamaño de la unidad:
1 miligramo
25 microgramos
50 microgramos
500 microgramos
Código de producto. Cantidad unitSize
17885113 25 μg 25 microgramos
17875653 50 μg 50 microgramos
17815313 2 x 500 μg 1 miligramo
30054491 500 μg 500 microgramos
4 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17885113

Marca: PeproTech 500P260BT25UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: VHHPPSYVAH LASDFGVRVF QQVAQASKDR NVVFSPYGVA SVLAMLQLTT GGETQQQIQA AMGFKIDDKG MAPALRHLYK ELMGPWNKDE ISTTDAIFVQ RDLKLVQGFM PHFFRLFRST VKQVDFSEVE RARFIINDWV KTHTKGMISN LLGKGAVDQL TRLVLVNALY FNGQWKTPFP DSSTHRRLFH KSDGSTVSVP MMAQTNKFNY TEFTTPDGHY YDILELPYHG DTLSMFIAAP YEKEVPLSAL TNILSAQLIS HWKGNMTRLP RLLVLPKFSL ETEVDLRKPL ENLGMTDMFR QFQADFTSLS DQEPLHVAQA LQKVKIEVNE SGTVASSSTA VIVSARMAPE EIIMDRPFLF VVRHNPTGTV LFMGQVMEP. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human PAI-1. Anti-Human PAI-1-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hPAI-1 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human PAI-1 (500-P260) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hPAI-1. Western Blot: To detect hPAI-1 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hPAI-1 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P260BT-1MG will be provided as 2 x 500 μg

PAI1 (plasminogen activator inhibitor 1) belongs to serine protease inhibitor superfamily, and is the principal inhibitor of tissue-type and urokinase-type plasminogen activators (tPA and uPA). Platelets are the main source of the circulating PAI11, but it is synthesized and secreted by many tissue and cell types, including fibroblasts, smooth muscle cells, endothelial cells, hepatocytes, and inflammatory cells. Expression of PAI11 can be regulated at the transcriptional level by many factors including growth factors, cytokines, hormones, inflammatory factors, glucose or lipid metabolites, vascular tone regulating factors, chemicals, and other environmental or physical factors. PAI1 is present at increased levels in various disease states, and has been linked to an increased occurrence of thrombosis in obesity, thrombophilia and the metabolic syndrome. Defects in PAI-1 are characterized by abnormal bleeding. PAI1 mediates inhibition of fibrinolysis by inhibiting the activity of plasminogen activator, and may promote neuronal survival. Other defects in PAI1 are the cause of plasminogen activator inhibitor-1 deficiency (PAI-1 deficiency). Alternatively spliced transcript variants encoding different isoforms of PAI1 have been found.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno PAI1
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Concentración 0.1-1.0 mg/mL
Conjugado Biotin
Formulación PBS with no preservative
génica Serpine1
N.º de referencia del gen P05121
Alias de gen Endothelial plasminogen activator inhibitor; nexin, plasminogen activator inhibitor type 1; PAI; PAI1; PAI-1; PAI1 protein; PAI1A; Pai1aa; Planh; Planh1; plas; plasminogen activator inhibitor; plasminogen activator inhibitor 1; plasminogen activator inhibitor I; plasminogen activator inhibitor, type I; plasminogen activator inhibitor-1; Plasminogen activator inhibitor-1 precursor (PAI-1) (Endothelial plasminogen activator inhibitor) (PAI); putative plasminogen activator inhibitor-1; RATPAI1A; serine (or cysteine) peptidase inhibitor, clade E, member 1; serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 1; serine (or cysteine) proteinase inhibitor, clade E, member 1; serine (or cysteine) proteinase inhibitor, member 1; serpin E1; serpin family E member 1; serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 1; SERPINE1; SERPINE-1
Símbolos de los genes Serpine1
Especie del huésped Rabbit
Inmunógeno E.coli-derived, 42.7 kDa Recombinant Human PAI-1.
Método de purificación Antigen affinity chromatography
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 5054
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Lyophilized
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.