missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAGE1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA542815
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Peptide sequence: TKRVCLRNEE QMKLPAEGPE PEADSQEQVH PKTGCERGDG PDVQELGLPN Sequence homology: Human: 100%.
PAGE1 belongs to a family that are expressed in a variety of tumors but not in normal tissues, except for the testis. Nothing is presently known about the function of this protein.This gene belongs to a family of genes that are expressed in a variety of tumors but not in normal tissues, except for the testis. Unlike the other gene family members, this gene does not encode an antigenic peptide. Nothing is presently known about the function of this protein.
Especificaciones
| PAGE1 | |
| Polyclonal | |
| Unconjugated | |
| PAGE1 | |
| AL5; CT16.3; G antigen 9; G antigen family B member 1; G antigen, family B, 1 (prostate associated); GAGE9; GAGE-9; GAGEB1; P antigen family member 1; P antigen family, member 1 (prostate associated); PAGE family member 1; PAGE1; PAGE-1; prostate associated gene 1; prostate-associated gene 1 protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 8712 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O75459 | |
| PAGE1 | |
| Synthetic peptide directed towards the middle region of human PAGE1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido