Learn More
PADI4 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Código de producto. 18084745 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
100 μL
20 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18084745 100 μL 100 microlitros
18018705 20 μL 20 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18084745 Proveedor Proteintech N.º de proveedor CL48817373100UL
Rabbit Polyclonal Antibody
The PADI4 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PADI4. This antibody recognizes human, mouse, rat antigen. The PADI4 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Especificaciones
Antígeno PADI4
Aplicaciones Immunofluorescence
Concentración 1000 μg/mL
Conjugado CoraLite Plus 488
Símbolos de los genes HL 60 PAD, PAD, PAD4, PADI4, PADI5, PDI4, PDI5, Peptidylarginine deiminase IV
Especie del huésped Rabbit
Cantidad 100 μL
Primario o secundario Primary
ID de gen (Entrez) 23569
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Peso molecular del anticuerpo 150 kDa
Formulario Liquid
Isotype IgG