missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PACT Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA565135
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: KLAKHRAAEAA INILKANASI CFAVPDPLMP DPSKQPKNQL NPIGSLQELA IHHGWRLPEY TLSQEGGPAH KREYTTICRL ESFMETGK Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
PRKRA appears to have a pro-apoptotic function that may be suppressed in the presence of growth factor. It activates EIF2AK2 in the absence of double-stranded RNA (dsRNA).
Especificaciones
| PACT | |
| Polyclonal | |
| Unconjugated | |
| PRKRA | |
| AV120107; DYT16; HSD14; HSD-14; interferon-inducible double stranded RNA-dependent protein kinase activator A; interferon-inducible double-stranded RNA-dependent protein kinase activator A; lear; PACT; PKR associated protein X; PKR-associated protein X; PKR-associating protein X; PRK; PRKRA; protein activator of interferon induced protein kinase EIF2AK2; protein activator of the interferon-induced protein kinase; protein kinase, interferon inducible double stranded RNA dependent activator; protein kinase, interferon-inducible double stranded RNA dependent activator; protein kinase, interferon-inducible double stranded RNA-dependent activator; protein kinase, interferon-inducible double-stranded RNA-dependent activator; RAX | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8575 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75569 | |
| PRKRA | |
| Recombinant Human PACT. Recombinant protein control fragment (Product #RP-104651). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido