missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PABP Polyclonal Antibody

Descripción
Immunogen sequence: VTEMNGRIVA TKPLYVALAQ RKEERQAHLT NQYMQRMASV RAVPNPVINP YQPAPPSGYF MAAIPQTQNR AAYYPPSQIA QLRPSPRWTA QGARPHPFQN M; Positive Samples: A-549, HT-29, HeLa, Mouse brain, Rat lung; Cellular Location: Cytoplasm, Nucleus.
The poly(A)-binding protein (PABP), which is found complexed to the 3-prime poly(A) tail of eukaryotic mRNA, is required for poly(A) shortening and translation initiation. In humans, the PABPs comprise a small nuclear isoform and a conserved gene family that displays at least 3 functional proteins: PABP1 (PABPC1), inducible PABP (iPABP, or PABPC4), and PABP3 (PABPC3). In addition, there are at least 4 pseudogenes, PABPCP1 to PABPCP4.
Especificaciones
Especificaciones
| Antígeno | PABP |
| Aplicaciones | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 1.88 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| génica | PABPC1 |
| N.º de referencia del gen | P11940, P29341, Q9EPH8 |
| Alias de gen | Cytoplasmic poly(A)-binding protein 1-A; ePAB; hypothetical protein; PAB1; pabp; PABP 1; Pabp1; PABP-1; PABP-1A; PABP-1-A; PABP-A; Pabpc1; pabpc1.L; pabpc1.S; pabpc1a; pabpc1-a; pabpc1a protein; pabpc1-b; pabpc2; PabpI; PABPL1; poly A binding protein 1; poly A binding protein, cytoplasmic 1 a; poly(A) binding protein cytoplasmic 1; poly(A) binding protein, cytoplasmic 1; poly(A) binding protein, cytoplasmic 1 L homeolog; poly(A) binding protein, cytoplasmic 1 S homeolog; poly(A) binding protein, cytoplasmic 1a; poly(A) binding protein, cytoplasmic 2; poly(A)-binding protein 1; poly(A)-binding protein 1A; poly(A)-binding protein 1-A; polyadenylate-binding protein 1; Polyadenylate-binding protein 1A; Polyadenylate-binding protein 1-A; polyadenylate-binding protein 1A; poly(A) binding protein, cytoplasmic 1a; wu:fb16a02; wu:fi19b08; wu:fj12d09; wu:fj61f06; XELAEV_18033915mg; xPABP1-A; zgc:109879 |
| Símbolos de los genes | PABPC1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?