missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p63/TP73L Antibody (TP63/2428), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP3-08736AF350
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
p63/TP73L Monoclonal antibody specifically detects p63/TP73L in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry (Paraffin), Protein Array
Especificaciones
| p63/TP73L | |
| Monoclonal | |
| Alexa Fluor 350 | |
| amplified in squamous cell carcinoma, Chronic μgcerative stomatitis protein, CUSP, EEC3B(p51B), Keratinocyte transcription factor KET, KETRHS, LMS, OFC8SHFM4TP73LNBP, p40, p51tumor protein p53-like, p53CP, P63, p63AIS, p73H, P73L, p73LB(p51A), TP53CP, TP53L, Transformation-related protein 63, tumor protein 63, tumor protein p53-competing protein, tumor protein p63, tumor protein p63 deltaN isoform delta, Tumor protein p73-like | |
| Recombinant full-length human p63/TP73L protein (Uniprot: Q9H3D4 ) | |
| 100 μg | |
| Apoptosis, Cancer, Cellular Markers, Hypoxia, Stem Cell Markers, Tumor Suppressors | |
| 8626 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Flow Cytometry, ELISA, Immunohistochemistry (Paraffin), Protein Array | |
| TP63/2428 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido