Learn More
p63 (Squamous, Basal & Myoepithelial Cell Marker) Mouse, Clone: TP63/2427, enQuireBio™
Mouse Monoclonal Antibody
Marca:  EnquireBio 8626-MSM7-P1
 Ver más versiones de este producto
Código de producto. 15951138
  • 528.00€ / 100 microgramos
Especificaciones
p63 (Squamous, Basal & Myoepithelial Cell Marker)
Nuclear
TP63/2427
Unconjugated
Mouse
63kDa
Protein A/G purified from Bioreactor concentrate.
RUO
8626
IgG2b κ
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Recombinant full-length human p63 protein
HEK293 cells or Prostate Carcinoma or Lung or bladder squamous cell carcinoma
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Certificados
Promociones
Promociones
Título del producto
p63 (Squamous, Basal & Myoepithelial Cell Marker) Mouse, Clone: TP63/2427, enQuireBio™ > 100 μg