missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ p38 MAPK gamma/delta Polyclonal Antibody

Description
Antibody detects endogenous levels of total p38 gamma/delta.
Activation of members of the mitogen-activated protein kinase family is a major mechanism for transduction of extracellular signals. Stress-activated protein kinases are one subclass of MAP kinases. The protein encoded by this gene functions as a signal transducer during differentiation of myoblasts to myotubes.
Specifications
Specifications
| Antígeno | p38 MAPK gamma/delta |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| génica | MAPK13 |
| N.º de referencia del gen | O08911, O15264, P53778, Q9Z1B7 |
| Alias de gen | AW123708; ERK3; ERK6; ERK-6; Exon skip; extracellular signal-regulated kinase 6; MAP kinase 12; MAP kinase 13; MAP kinase p38 delta; MAP kinase p38 gamma; MAPK 12; MAPK 13; MAPK12; MAPK13; MAPK-13; mitogen activated protein kinase 12; mitogen activated protein kinase 13; mitogen-activated protein kinase 12; Mitogen-activated protein kinase 13; mitogen-activated protein kinase 3; mitogen-activated protein kinase p38 delta; Mitogen-activated protein kinase p38 gamma; p38 delta MAP kinase; p38delta; P38GAMMA; PRKM12; Prkm13; SAP kinase-3; SAPK/Erk/kinase 4; SAPK3; SAPK-3; SAPK4; Serk4; stress activated protein kinase 3; stress-activated protein kinase 3; Stress-activated protein kinase 4 |
| Símbolos de los genes | MAPK12, MAPK13 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?