Description
p19 / INK 4d is a specific inhibitor of cdk 4 / cdk 6 and is periodically expressed during cell cycle. It is maximally induced as cells enter S-phase.
Specifications
Specifications
| Antígeno | p19 INK4d |
| Aplicaciones | Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence |
| Clasificación | Polyclonal |
| Concentración | 0.23 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | CDKN2D |
| N.º de referencia del gen | P55273, Q60773 |
| Alias de gen | CDKN2D, INK4D, P19, p19 INK4D, p19 INK4d |
| Símbolos de los genes | Cdkn2d |
| Show More |