missing translation for 'onlineSavingsMsg'
Learn More

p16INK4a/CDKN2A Antibody (CDKN2A/7660R) [Janelia Fluor™ 635], Novus Biologicals™

Código de producto. 30172773
Change view
missing translation for 'orderingAttributeHoverText'
Cantidad:
0.1 mL
missing translation for 'unitSize'
0.10 ml
Product Code. Cantidad unitSize
30172773 0.1 mL 0.10 ml
1 missing translation for 'options'
This item is not returnable. View return policy

Product Code. 30172773

missing translation for 'mfr': Novus Biologicals (Bio-Techne) NBP320900JF635

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

p16INK4a/CDKN2A Monoclonal antibody specifically detects p16INK4a/CDKN2A in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno p16INK4a/CDKN2A
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon CDKN2A/7660R
Conjugado Janelia Fluor 635
Formulación 50mM Sodium Borate
Alias de gen ARF, CDK4 inhibitor p16-INK4, CDK4IP14ARF, CDKN2, cell cycle negative regulator beta, CMM2P16-INK4A, Cyclin-dependent kinase 4 inhibitor A, cyclin-dependent kinase inhibitor 2A, cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4), INK4, INK4A, MLMP16INK4, MTS-1, MTS1P14, Multiple tumor suppressor 1, p14, p14ARF, p16, p16-INK4, p16INK4a, p16-INK4a, P19, p19ARF, TP16
Especie del huésped Rabbit
Inmunógeno Purified recombinant prokaryotic full-length human p16INK4a/CDKN2A protein
Método de purificación Protein A or G purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación ABC Transporters, Biologically Active Proteins, Cancer, Cell Cycle and Replication, DNA Repair, Hypoxia, MAP Kinase Signaling, Neuroscience, Tumor Suppressors
Primario o secundario Primary
ID de gen (Entrez) 1029
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.