Learn More
p150 glued Rabbit anti-Canine, Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16856665 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16856665 20 μL 20 microlitros
16846665 150 μL 150 microlitros
2 options
Código de producto. 16856665
Marca: Proteintech 551821AP20UL
Rabbit Polyclonal Antibody
This antibody is specific to DCTN1.
This gene encodes the largest subunit of dynactin, a macromolecular complex consisting of 10 subunits ranging in size from 22 to 150 kD. Dynactin binds to both microtubules and cytoplasmic dynein. Dynactin is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit interacts with dynein intermediate chain by its domains directly binding to dynein and binds to microtubules via a highly conserved glycine-rich cytoskeleton-associated protein (CAP-Gly) domain in its N-terminus. Alternative splicing of this gene results in multiple transcript variants encoding distinct isoforms. Mutations in this gene cause distal hereditary motor neuronopathy type VIIB (HMN7B) which is also known as distal spinal and bulbar muscular atrophy (dSBMA).
Especificaciones
Antígeno p150 glued
Aplicaciones Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence
Clasificación Polyclonal
Concentración 0.17 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica DCTN1
N.º de referencia del gen O08788, P28023, Q14203
Alias de gen DAP 150, DCTN1, DP 150, Dynactin subunit 1, HMN7B, P135, p150 glued
Símbolos de los genes DCTN1
Especie del huésped Rabbit
Inmunógeno Peptide
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 13191, 1639, 29167, 483106
Especies diana Rat, Mouse, Human, Canine
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG