Learn More
P glycoprotein Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Código de producto. 16881865 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16881865 150 μL 150 microlitros
16891865 20 μL 20 microlitros
2 options
Código de producto. 16881865
Marca: Proteintech 223361AP150UL
Rabbit Polyclonal Antibody
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The protein encoded by this gene is an ATP-dependent drug efflux pump for xenobiotic compounds with broad substrate specificity. It is responsible for decreased drug accumulation in multidrug-resistant cells and often mediates the development of resistance to anticancer drugs. This protein also functions as a transporter in the blood-brain barrier.
Especificaciones
Antígeno P glycoprotein
Aplicaciones Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Clasificación Polyclonal
Concentración 0.48 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica ABCB1
N.º de referencia del gen P06795, P08183, P21447
Alias de gen ABCB1, CD243, GP170, MDR1, Multidrug resistance protein 1, P glycoprotein 1
Símbolos de los genes ABCB1, Abcb1a, Abcb1b
Especie del huésped Rabbit
Inmunógeno P glycoprotein Fusion Protein Ag17811
Método de purificación Antigen Affinity Chromatography
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 18669, 18671, 5243
Especies diana Human, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG