missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Oxytocin Polyclonal Antibody
GREENER_CHOICE

Código de producto. 17711744
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
17711744 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17711744

Marca: Invitrogen™ PA5120341

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Positive test controls include: A549, NCI-H460, LO2, HepG2, Mouse liver, Rat liver, Rat lung. The target is usually found in the following locations: Secreted. Immunogen sequence: MAGPSLACCL LGLLALTSAC YIQNCPLGGK RAAPDLDVRK CLPCGPGGKG RCFGPNICCA EELGCFVGTA EALRCQEENY LPSPCQSGQK ACGSGGRCAV LGLCCSPDGC HADPACDAEA TFSQR.

There are two proteins encoded by this gene, oxytocin and neurophysin I. Oxytocin is posterior pituitary hormone which is synthesized as an inactive precursor in the hypothalamus along with its carrier protein neurophysin I. Together with neurophysin, it is packaged into neurosecretory vesicles and transported axonally to the nerve endings in the neurohypophysis, where it is either stored or secreted into the bloodstream. The precursor seems to be activated while it is being transported along the axon to the posterior pituitary. This hormone contracts smooth muscle during parturition and lactation. It is also involved in cognition, tolerance, adaptation and complex sexual and maternal behaviour, as well as in the regulation of water excretion and cardiovascular functions.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Oxytocin
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Concentración 1.95 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
génica OXT
N.º de referencia del gen P01178, P01179, P35454
Alias de gen neurophysin 1; neurophysin I; Ocytocin; Ot; OT-NPI; oxcytocin; OXT; OXT-NPI; Oxy; Oxytocin; oxytocin, prepro- (neurophysin I); oxytocin, prepropeptide; oxytocin/neurophysin (Oxt) gene, complete gene, complete cds; oxytocin/neurophysin 1 prepropeptide; oxytocin/neurophysin I prepropeptide; oxytocin-neurophysin 1; oxytocin-neurophysin I; oxytocin-neurophysin I, preproprotein; prepropeptide for oxytocin and neurophysin; unnamed protein product
Símbolos de los genes OXT
Especie del huésped Rabbit
Inmunógeno Recombinant fusion protein containing a sequence corresponding to amino acids 1-125 of human OXT (NP_000906.1).
Método de purificación Affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 18429, 25504, 5020
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.