Learn More
Thermo Scientific™ Medios en frasco de caldo de enriquecimiento para Salmonella RV preparados Oxoid™
Código de producto. 12583946
Change view
Click to view available options
:
Volumen: 10 ml
Código de producto.
12583946 Volumen: 10 ml
1 options
Código de producto. 12583946
Marca: Thermo Scientific™ BO1121E
Selectively enrich for the isolation of salmonellae from food and environmental specimens with Thermo Scientific™ Rappaport-Vassiliadis Soya Peptone Broth prepared media tubes. RVS Peptone Broth exploits characteristics of Salmonella species when compared with other Enterobacteriaceae, such as the ability to survive at relatively high osmotic pressure, multiply at relatively low pH values, be relatively more resistant to malachite green, and have relatively less demanding nutritional requirements.