missing translation for 'onlineSavingsMsg'
Learn More

OX40/TNFRSF4 Antibody (tavolixizumab) [Janelia Fluor™ 635], Novus Biologicals™

Código de producto. 30177627
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30177627 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Product Code. 30177627 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP328570JF635

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Human Monoclonal Antibody

OX40/TNFRSF4 Monoclonal antibody specifically detects OX40/TNFRSF4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antígeno OX40/TNFRSF4
Aplicaciones Flow Cytometry, ELISA, Functional Assay
Clasificación Monoclonal
Clon tavolixizumab
Conjugado Janelia Fluor 635
Formulación 50mM Sodium Borate
Alias de gen ACT35 antigen, ACT35ATC35 antigen, CD134, CD134 antigen, OX40 cell surface antigen, OX40 homologue, OX40L receptor, OX40lymphoid activation antigene ACT35, TAX transcriptionally-activated glycoprotein 1 receptor, tax-transcriptionally activated glycoprotein 1 receptor, tumor necrosis factor receptor superfamily member 4, tumor necrosis factor receptor superfamily, member 4, TXGP1LOX40 antigen
Especie del huésped Human
Inmunógeno TNFRSF4/ OX40/ CD134
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cell Cycle and Replication, Immunology, Myeloid derived Suppressor Cell, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 7293
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.