Learn More
Invitrogen™ OX40 Monoclonal Antibody (OX-86), PE
Descripción
OX40 Monoclonal Antibody for Flow
OX40 is a protein receptor found on the surface of T cells, a type of white blood cell that plays a crucial role in the immune system. When activated, OX40 promotes the proliferation and survival of T cells, leading to a stronger immune response against cancer cells and infectious agents. OX40 agonists, which are drugs that activate OX40, have shown promising results in preclinical and clinical studies as a potential immunotherapy for cancer. They can enhance the efficacy of other cancer treatments such as chemotherapy and checkpoint inhibitors, and may also have applications in autoimmune diseases and allergies.
Especificaciones
Especificaciones
| Antígeno | OX40 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | OX-86 |
| Concentración | 0.1 mg/mL |
| Conjugado | PE |
| Formulación | PBS with 4-5mg/mL BSA and 0.02% sodium azide |
| génica | Tnfrsf4 |
| N.º de referencia del gen | P47741 |
| Alias de gen | ACT35; ACT35 antigen; ATC35 antigen; CD134; CD134 antigen; IMD16; Ly-70; lymphoid activation antigene ACT35; MRC OX40; Ox40; OX40 antigen; OX40 cell surface antigen; OX40 homologue; OX40L receptor; TAX transcriptionally-activated glycoprotein 1 receptor; tax-transcriptionally activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 receptor; TNF receptor superfamily member 4; Tnfrsf4; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor receptor superfamily member 4; tumor necrosis factor receptor superfamily, member 4; tumor necrosis factor superfamily, member 4; Txgp1; Txgp1l |
| Mostrar más |