missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Otubain-1 Antibody, CoraFluor™ 1, Novus Biologicals™
Description
Otubain-1 Polyclonal antibody specifically detects Otubain-1 in Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antígeno | Otubain-1 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Conjugado | CoraFluor 1 |
| Formulación | PBS |
| Alias de gen | Deubiquitinating enzyme OTUB1, EC 3.1.2, EC 3.4.19.12, FLJ20113, FLJ40710, hOTU1, MGC111158, OTB1MGC4584, OTU domain, ubiquitin aldehyde binding 1, OTU domain-containing ubiquitin aldehyde-binding protein 1, OTU1OTU-domain Ubal-binding 1, otubain-1, ubiquitin thioesterase OTUB1, ubiquitin-specific protease otubain 1, Ubiquitin-specific-processing protease OTUB1 |
| Especie del huésped | Rabbit |
| Inmunógeno | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Otubain-1 |
| Método de purificación | Antigen and protein A Affinity-purified |
| Cantidad | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?