missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OSCAR Monoclonal Antibody (C2), Invitrogen™

Descripción
Recently, a novel member of the leukocyte receptor complex (LRC) encoded family has been identified that is expressed specifically in osteoclast (OC). This protein names OC-associated receptor or OSCAR, play critical roles in the regulation of both innate and adaptive immune responses and bone-specific regulator of OC differentiation. Osteoclasts are multinucleated cells that resorb bone and are essential for bone homeostasis. This protein family plays critical roles in the regulation of both innate and adaptive immune responses. Different from the other LRC members, OSCAR expression is detected specifically in preosteoclasts or mature osteoclasts. OSCAR may be an important bone-specific regulator of osteoclast differentiation. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Especificaciones
Especificaciones
| Antígeno | OSCAR |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | C2 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| génica | OSCAR |
| N.º de referencia del gen | Q8IYS5 |
| Alias de gen | MGC33613; mOSCAR; mOSCAR-M1; mOSCAR-M2; mOSCAR-M3; ocl178; Oscar; osteoclast associated receptor; osteoclast associated receptor OSCAR-S1; osteoclast associated receptor OSCAR-S2; osteoclast associated, immunoglobulin-like receptor; osteoclast-associated immunoglobulin-like receptor; Osteoclast-associated receptor; osteoclast-associated receptor OSCAR; PIGR3; PIgR-3; poly-Ig receptor 3; polymeric immunoglobulin receptor 3 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?