missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OS9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA542628
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Peptide sequence: LKEIFFNILV PGAEEAQKER QRQKELESNY RRVWGSPGGE GTGDLDEFDF Sequence homology: Cow: 100%; Dog: 86%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%.
OS9 is an ER glycoprotein found in the intralumenal level and highly expressed in tumor tissue. The OS9 gene is transcriptionally induced upon activation of the Ire1/Xbp1 ER-stress pathway.
Especificaciones
| OS9 | |
| Polyclonal | |
| Unconjugated | |
| OS9 | |
| 4632413K17Rik; amplified in osteosarcoma; amplified in osteosarcoma 9; AU022351; endoplasmic reticulum lectin 2; ERLEC2; erlectin 2; OS9; Os-9; OS9 endoplasmic reticulum lectin; OS9, endoplasmic reticulum lectin; osteosarcoma amplified 9; osteosarcoma amplified 9, endoplasmic reticulum associated protein; osteosarcoma amplified 9, endoplasmic reticulum lectin; protein OS-9 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10956 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q13438 | |
| OS9 | |
| Synthetic peptide directed towards the C-terminal of human OS9 (aa 618-667). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido