missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORM2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Especificaciones
| Antígeno | ORM2 |
|---|---|
| Concentración | 0.45 mg/mL |
| Aplicaciones | Immunoprecipitation, Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
|
16808123
|
Proteintech
11199-1-AP-20UL |
20 μL |
145.00€
20 microlitros |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
|
16898113
|
Proteintech
11199-1-AP-150UL |
150 μL |
410.00€
150 microlitros |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
This antibody can recognize both ORM1 and ORM2.
AGP (a 1-acid Glycoprotein) is an acute phase plasma protein synthesized by the liver. It functions to regulate the interaction between blood cells and endothelial cells, and together with haptoglobin and C reactive protein, it also mediates the extravasation of cells during infection and inflammation. Expression of AGP is induced by acute-phase stimulatory agents such as bacterial lipopolysaccharides. AGP has a high affinity, low capacity binding for basic drugs at physiological pH. In human plasma, AGP is found at levels of 0.5-1.4 mg/ml, though this is elevated during acute inflammation, and, as a result, levels of this protein can be used to diagnose inflammatory conditions. Multiple AGP genes exist, including AGP-1, AGP-2, AGP-3 and AGP-8. AGP-1 and AGP-2 contain five and six potential N-glycosylation sites, respectively. Abnormal expression of the APG-1 gene is linked to sarcoidosis and other immunogenetic diseases, while mutations in the APG-2 gene are associated with different types of carcinomas.Especificaciones
| ORM2 | |
| Immunoprecipitation, Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P07361, P19652 | |
| 18406, 5005 | |
| ORM2 Fusion Protein Ag1667 | |
| Primary | |
| -20°C | |
| ORM2 |
| 0.45 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| AGP 2, AGP B, AGP B′, AGP2, Alpha 1 acid glycoprotein 2, OMD 2, ORM2, orosomucoid 2 | |
| Orm2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto