missing translation for 'onlineSavingsMsg'
Learn More

ONO-4059 analog, MedChemExpress

Código de producto. 16433667
Encompass_Preferred
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
25 miligramos
5 miligramos
50 miligramos
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16433667

Marca: Medchem Express HY18951

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

MedChemExpress ONO-4059 analog is the analog of ONO-4059, ONO-4059 is a highly potent and selective Btk inhibitor.

ONO-4059 analog is the analog of ONO-4059, ONO-4059 is a highly potent and selective Btk inhibitor.

Specifications

Nombre del producto químico o material ONO-4059 analog
CAS 1351635-67-0
Color Off-White
Notas de grado de pureza Research
Fórmula molecular C25H24N6O3
Cantidad 50 mg
Información de solubilidad DMSO : ≥ 29 mg/mL (63.53 mM)
SMILES O=C1N([C@H]2CCCN(C(C=C)=O)C2)C3=NC=NC(N)=C3N1C4=CC=C(OC5=CC=CC=C5)C=C4
Peso molecular (g/mol) 456.5
Formula Weight (peso de la fórmula) 456.5
Porcentaje de pureza 98.0%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.