missing translation for 'onlineSavingsMsg'
Learn More

Ombrabulin hydrochloride, MedChemExpress

Product Code. 16479967
Encompass_Preferred
Change view
Click to view available options
Cantidad:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
1 miligramo
10 miligramos
100 miligramos
5 miligramos
50 miligramos
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
16479967 5 mg 5 miligramos
16497446 100 mg 100 miligramos
16457488 10 mg 10 miligramos
16431729 1 mg 1 miligramo
16403517 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16479967 Supplier Medchem Express Supplier No. HY18256

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

MedChemExpress Ombrabulin hydrochloride is a derivative of CA-4 phosphate, which is known to exhibit antivascular effects through selective disruption of the tubulin cytoskeleton of endothelial cells.

Ombrabulin hydrochloride is a derivative of CA-4 phosphate, which is known to exhibit antivascular effects through selective disruption of the tubulin cytoskeleton of endothelial cells.

Specifications

Nombre del producto químico o material Ombrabulin hydrochloride
CAS 253426-24-3
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C21H27ClN2O6
Cantidad 5 mg
Sinónimo AVE8062 hydrochlorideAC7700 hydrochloride
Información de solubilidad H2O : 20 mg/mL (45.57 mM; Need ultrasonic)
SMILES O=C(NC1=CC(/C=C\C2=CC(OC)=C(OC)C(OC)=C2)=CC=C1OC)[C@@H](N)CO.[H]Cl
Peso molecular (g/mol) 438.9
Formula Weight (peso de la fórmula) 438.9
Porcentaje de pureza 99.43%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.