missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OGFOD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA569640
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 93%.
OGFOD3 gene ontology annotations related to this gene include dioxygenase activity; iron ion binding; L-ascorbic acid binding; oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen.
Especificaciones
| OGFOD3 | |
| Polyclonal | |
| Unconjugated | |
| OGFOD3 | |
| 1110031I02Rik; 2-oxoglutarate and iron dependent oxygenase domain containing 3; 2-oxoglutarate and iron-dependent oxygenase domain containing 3; 2-oxoglutarate and iron-dependent oxygenase domain-containing protein 3; C17orf101; Ogfod3; PKHD domain-containing transmembrane protein C17orf101; PKHD domain-containing transmembrane protein C17orf101 homolog; RGD1305007 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 303749 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5M843 | |
| OGFOD3 | |
| synthetic peptide directed towards the N-terminal region of rat OGFOD3. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido