Learn More
OGDH Mouse anti-Human, CoraLite 488, Clone: 1H7A2, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL488-66285-100UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The OGDH antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human OGDH. This antibody recognizes human, rat antigen. The OGDH antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Especificaciones
| OGDH | |
| Monoclonal | |
| CoraLite 488 | |
| 100 μL | |
| 4967 | |
| IgG2b |
| Immunofluorescence | |
| 1H7A2 | |
| Mouse | |
| Primary | |
| Human, Rat |