missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCT4 (OCT3) Mouse, anti-Human, Unconjugated, Clone: 3A2A20, STEMCELL Technologies
Descripción
- The 3A2A20 antibody reacts with human OCT4 (octamer-binding transcription factor 4; also known as OCT3 and OCT3/4), an ∼40 kDa homeodomain transcription factor belonging to the POU family, which is expressed in undifferentiated human embryonic stem (ES), induced pluripotent stem (iPS), embryonal carcinoma (EC) and embryonic germ (EG) cells
- OCT4 binds to the octamer motif (5'-ATTTGCAT-3') and plays a key role in maintaining cells in a pluripotent state by interacting with other transcription factors such as SOX2 to regulate the expression of several genes, including FBX15, FGF-4, REX1, SOX2 and osteopontin
- Levels of OCT4 are down-regulated during differentiation and it has thus emerged as a useful marker of pluripotency in stem cells, as well as a marker for certain human malignant germ cell tumours
- Expression of OCT4 together with other transcription factors has been used to reprogram somatic cells into iPS cells
Especificaciones
Especificaciones
| Antígeno | OCT4 (OCT3) |
| Aplicaciones | Flow Cytometry, Western Blot, Immunocytochemistry, Immunofluorescence |
| Clasificación | Monoclonal |
| Clon | 3A2A20 |
| Conjugado | Unconjugated |
| Especie del huésped | Mouse |
| Cantidad | 25 μg |
| Primario o secundario | Primary |
| Especies diana | Human |
| Isotype | IgG2b κ |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?