Learn More
Occludin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Código de producto. p-7211625 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16805525 20 μL 20 microlitros
16895515 150 μL 150 microlitros
2 options
Código de producto. 16805525
Marca: Proteintech 272601AP20UL
Rabbit Polyclonal Antibody
This gene encodes an integral membrane protein that is required for cytokine-induced regulation of the tight junction paracellular permeability barrier. Mutations in this gene are thought to be a cause of band-like calcification with simplified gyration and polymicrogyria (BLC-PMG), an autosomal recessive neurologic disorder that is also known as pseudo-TORCH syndrome. Alternative splicing results in multiple transcript variants. A related pseudogene is present 1.5 Mb downstream on the q arm of chromosome 5.
Especificaciones
Antígeno Occludin
Aplicaciones Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 0.17 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica OCLN
N.º de referencia del gen Q16625, Q61146
Alias de gen Occludin, OCLN
Símbolos de los genes Ocln
Especie del huésped Rabbit
Inmunógeno Occludin Fusion Protein Ag26173
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 100506658, 18260
Especies diana Human, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG