Learn More
Invitrogen™ Nur77 Recombinant Rabbit Monoclonal Antibody (HL2660)
GREENER_CHOICE
Código de producto. 30276129
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
30276129 100 μL 100 microlitros
1 options
Código de producto. 30276129
Marca: Invitrogen™ MA556818
Rabbit Recombinant Monoclonal Antibody
Nur77 Recombinant Monoclonal Antibody for Western Blot
NR4A1 is a member of the steroid-thyroid hormone-retinoid receptor superfamily. NGFI-B-alpha is expressed in brain, adrenal, thyroid, liver, testis, ovary, thymus, muscle, lung and prostate. Its expression is induced in response to various stress stimuli and growth factors. The encoded protein acts as a nuclear transcription factor involved in mediating apoptotic signaling in thymocytes and tumor cells, as well as having a role in signaling in the hypothalamic-pituitary axis. Translocation of the protein from the nucleus to mitochondria induces apoptosis. Three transcript variants encoding two distinct isoforms have been identified for this NR4A1. Variant (3), also known as TRC beta, contains multiple differences compared to variant 1. The resulting isoform b contains a shorter and distinct C-terminus compared to isoform a. RXR has been shown to be a partner for NR4A1.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Nur77
Aplicaciones Western Blot
Clasificación Recombinant Monoclonal
Clon HL2660
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with no preservative
génica NR4A1
N.º de referencia del gen P22736
Alias de gen early response protein NAK1; Gfrp; GFRP1; growth factor-inducible nuclear protein N10; Hbr1; Hbr-1; Hmr; hormone receptor; immediate early gene transcription factor NGFI-B; MGC9485; N10; NAK1; NAK-1; nerve growth factor IB nuclear receptor variant 1; nerve growth factor induced protein I-B; nerve growth factor-induced protein I-B; Ngfib; NGFI-B; NP10; Nr4a1; Nuclear hormone receptor NUR/77; nuclear protein N10; Nuclear receptor subfamily 4 group A member 1; nuclear receptor subfamily 4, group A, member 1; NUR77; Orphan nuclear receptor HMR; orphan nuclear receptor TR3; Orphan receptor tr3; ST-59; steroid receptor TR3; testicular receptor 3; TIS1; TR3; TR3 orphan receptor
Símbolos de los genes NR4A1
Especie del huésped Rabbit
Inmunógeno Recombinant fragment of human NUR77.
Método de purificación Protein A
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3164
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG