missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
NR4A1 is a member of the steroid-thyroid hormone-retinoid receptor superfamily. NGFI-B-alpha is expressed in brain, adrenal, thyroid, liver, testis, ovary, thymus, muscle, lung and prostate. Its expression is induced in response to various stress stimuli and growth factors. The encoded protein acts as a nuclear transcription factor involved in mediating apoptotic signaling in thymocytes and tumor cells, as well as having a role in signaling in the hypothalamic-pituitary axis. Translocation of the protein from the nucleus to mitochondria induces apoptosis. Three transcript variants encoding two distinct isoforms have been identified for this NR4A1. Variant (3), also known as TRC beta, contains multiple differences compared to variant 1. The resulting isoform b contains a shorter and distinct C-terminus compared to isoform a. RXR has been shown to be a partner for NR4A1.
Especificaciones
Especificaciones
| Antígeno | NUR77 |
| Aplicaciones | Immunocytochemistry, Immunofluorescence, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.18 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | NR4A1 |
| N.º de referencia del gen | P12813, P22736, P22829 |
| Alias de gen | Early response protein NAK1, GFRP1, HMR, N10, NAK 1, NAK1, NGFIB, NR4A1, NUR77, Orphan nuclear receptor HMR, Orphan nuclear receptor TR3, ST 59, Testicular receptor 3, TR3 |
| Símbolos de los genes | NR4A1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?