Learn More
NUP153 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16882714 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16882714 150 μL 150 microlitros
16892714 20 μL 20 microlitros
2 options
Código de producto. 16882714
Marca: Proteintech 141891AP150UL
Rabbit Polyclonal Antibody
Nuclear pore complexes regulate the transport of macromolecules between the nucleus and cytoplasm. They are composed of at least 100 different polypeptide subunits, many of which belong to the nucleoporin family. Nucleoporins are glycoproteins found in nuclear pores and contain characteristic pentapeptide XFXFG repeats as well as O-linked N-acetylglucosamine residues oriented towards the cytoplasm. The protein encoded by this gene has three distinct domains: a N-terminal region containing a pore targeting and an RNA-binding domain , a central region containing multiple zinc finger motifs, and a C-terminal region containing multiple XFXFG repeats. Alternative splicing results in multiple transcript variants of this gene.
Especificaciones
Antígeno NUP153
Aplicaciones Immunoprecipitation, Western Blot
Clasificación Polyclonal
Concentración 0.17 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica NUP153
N.º de referencia del gen P49790, P49791
Alias de gen 153 kDa nucleoporin, HNUP153, N153, nucleoporin 153kDa, Nucleoporin Nup153, NUP153
Símbolos de los genes NUP153
Especie del huésped Rabbit
Inmunógeno NUP153 Fusion Protein Ag5519
Método de purificación Antigen Affinity Chromatography
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 218210, 25281, 9972
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG