Learn More
Thermo Scientific™ Nunc™ Multidishes with HydroCell™ Surface
Descripción
Inhibit the attachment of adherent cells and enable the formation of cell clusters with Thermo Scientific™ Nunc™ Multidishes with Thermo Scientific™ HydroCell™ Surface, featuring super-low cell binding capacity with premium quality.
A covalently immobilized super hydrophilic polymer forms a thin, even layer on cultureware with HydroCell Surface, resulting in minimal cell adhesion and protein adsorption. Depending on cell type, cells can be grown as a suspension of single cells or as a suspension of cell clusters.
- Minimal cell adhesion—high yield of cells and eliminate unwanted adhesion-induced differentiation
- Minimal protein adsorption—high yield of cell-secreted proteins
- Ideal for culturing embryroid bodies, neurospheres, or other anchorage-independent cell clusters
- Sterile and non-pyrogenic
- Customized bar coding is available
Ordering Information:
Individually wrapped with lid.
Especificaciones
Especificaciones
| Certificaciones/conformidad | Todas las microplacas fabricadas por Nunc cumplen la recomendación de las normas nacionales americanas (ANSI) (ANSI/SBS 1-2004) |
| Color del tapón | Clear |
| Cierre incluido | Tapón incluido |
| Forma del fondo | Plano |
| Color | Clear |
| Área de cultivo | 9.6 sq. cm |
| Esterilidad | Estéril |
| Tratamiento de superficie | HydroCell |
| Tipo | Multi Dish |
| Número de pocillos | 6 |
| Mostrar más |
For research and single-use only.
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.