Learn More
Thermo Scientific™ Matraces Nunc™ EasYFlask™ con superficie Nunclon™ Supra
Código de producto. 30408801
Click to view available options
Área de cultivo:
175 cm²
25 cm²
75 cm²
Tipo de tapón:
Filtro
Sólido
Tamaño de la unidad:
paquete de 100
paquete de 200
paquete de 30
Código de producto. 30408801
Marca: Thermo Scientific™ 156374
Experience optimal performance with Thermo Scientific Nunc EasYFlask flasks with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells.
Experience optimal performance with Thermo Scientific Nunc EasYFlask flasks with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells. The ergonomic design with angled neck and wide opening allows easy access to the growth surface by pipettes or cell scrapers. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
Thermo Scientific Nunc vessels with Nunclon Supra surface treatment are designed for culturing fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
The Nunclon Supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Nunclon Supra cultureware is a great xeno-free solution for cell therapy research applications.
Features of Nunc EasYFlask flasks with Nunclon Supra surface include:
• Consistent lot-to-lot performance
• Sterile with SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• Biocompatible per USP Class VI for raw materials
• Shelf life: 5 years
TRUSTED_SUSTAINABILITY
Especificaciones
Descripción Nunc EasYFlask, 25 cm²
N.º por caja 200 matraces
N.° por paquete 10 matraces
Tipo de tapón Sólido
Área de cultivo 25 cm²
Para uso en laboratorios