Learn More
Invitrogen™ NuMA Recombinant Rabbit Monoclonal Antibody (24GB1290)
GREENER_CHOICE
Código de producto. 30276617
Change view
Click to view available options
Cantidad:
20 μL
100 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
30276617 100 μL 100 microlitros
30274939 20 μL 20 microlitros
2 options
Código de producto. 30276617
Marca: Invitrogen™ MA557526
Rabbit Recombinant Monoclonal Antibody
NuMA Recombinant Monoclonal Antibody for Western Blot, Flow
NuMA (nuclear mitotic apparatus) is a long coil-coiled protein that plays a role in the interphase and mitosis phases of a cell cycle. In interphase, it has been hypothesized to play a structural role in the nucleoskeleton that may be important in nuclear organization and functions. During mitosis, it is associated with spindle microtubule organization and chromosome positioning.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno NuMA
Aplicaciones Flow Cytometry, Western Blot, Immunocytochemistry
Clasificación Recombinant Monoclonal
Clon 24GB1290
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
génica NUMA1
N.º de referencia del gen E9Q7G0, Q14980
Alias de gen 6720401E04Rik; AA764025; AL022610; AU014979; centrophilin stabilizes mitotic spindle in mitotic cells; NMP22; NMP-22; Nuclear matrix protein-22; Nuclear mitotic apparatus protein; Nuclear mitotic apparatus protein 1; NUMA; NuMA protein; Numa1; SP-H antigen; structural nuclear protein
Símbolos de los genes NUMA1
Especie del huésped Rabbit
Inmunógeno A synthesized peptide derived from human NUMA1 (aa 2000-2100).
Método de purificación Affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 101706, 308870, 4926
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG