missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUFIP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5113618
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: GQPWNFHAST SWYWRQSSDR FPRHQKSFNP AVKNSYYPRK YDAKFTDFSL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.
This gene encodes a nuclear RNA binding protein that contains a C2H2 zinc finger motif and a nuclear localization signal. This protein is associated with the nuclear matrix in perichromatin fibrils and, in neurons, localizes to the cytoplasm in association with endoplasmic reticulum ribosomes. This protein interacts with the fragile X mental retardation protein (FMRP), the tumor suppressor protein BRCA1, upregulates RNA polymerase II transcription, and is involved in box C/D snoRNP biogenesis. A pseudogene of this gene resides on chromosome 6q12.
Especificaciones
| NUFIP1 | |
| Polyclonal | |
| Unconjugated | |
| Nufip1 | |
| bA540M5.1; nuclear FMRP interacting protein; nuclear FMRP-interacting protein 1; nuclear fragile X mental retardation protein interacting protein 1; nuclear fragile X mental retardation-interacting protein 1; Nufip; NUFIP1; NUFIP1, FMR1 interacting protein 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 26747 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UHK0 | |
| Nufip1 | |
| Synthetic peptide directed towards the N terminal region of human NUFIP1 (aa 107-156). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido