Descripción
The NUDT13 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human NUDT13. This antibody recognizes human antigen. The NUDT13 antibody has been validated for the following applications: WB, ELISA analysis.
Especificaciones
Especificaciones
| Antígeno | NUDT13 |
| Aplicaciones | Western Blot, ELISA |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Especie del huésped | Rabbit |
| Cantidad | 20 μL |
| Primario o secundario | Primary |
| ID de gen (Entrez) | 25961 |
| Especies diana | Human |
| Isotype | IgG |