Learn More
Invitrogen™ Nuclear Matrix Protein p84 Recombinant Rabbit Monoclonal Antibody (24GB2205)
Descripción
Nuclear Matrix Protein p84 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
HPR1 is part of the TREX (transcription/export) complex, which includes TEX1 (MIM 606929), THO2 (MIM 300395), ALY (MIM 604171), and UAP56 (MIM 142560).
Especificaciones
Especificaciones
| Antígeno | Nuclear Matrix Protein p84 |
| Aplicaciones | Flow Cytometry, Western Blot, Immunocytochemistry |
| Clasificación | Recombinant Monoclonal |
| Clon | 24GB2205 |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| génica | Thoc1 |
| N.º de referencia del gen | P59924, Q8R3N6, Q96FV9 |
| Alias de gen | 3110002N20Rik; AW107452; HPR1; hTREX84; I79_014746; LRRGT00070; NMP-84; Nuclear matrix protein p84; P84; P84N5; THO complex 1; THO complex subunit 1; THO complex subunit 1-like protein; tho1; Thoc1 |
| Símbolos de los genes | Thoc1 |
| Mostrar más |