missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NTHL1 Recombinant Rabbit Monoclonal Antibody (8P6Z4)
Descripción
Immunogen Sequence: MCSPQESGMT ALSARMLTRS RSLGPGAGPR GCREEPGPLR RREAAAEARK SHSPVKRPRK AQRLRVAYEG SDSEKGEGAE PLKVPVWEPQ DWQQQLVNIR AMRNKKDAPV DHLGTEHCYD SSAPPKVRRY QVLLSL.
Oxygen radicals damage chromosonal DNA causing cell death and inducing mutations. Although a major focus of oxidatively damaged DNA has centered on the repair of 8-oxo-G, a number of other damaged DNA sites are created by free-radical attack on DNA. The human NTH1 repair protein is one enzyme that has been shown to act on a large number of these other oxidatively damaged DNA sites.A homologue of E. coli Endonuclease III, NTH is a DNA glycosylase with apurinic/apyrimidinic lyase activity. The NTH protein has broad substrate specificity, including numerous ring saturation and fragmentation products of pyrimidines. Research indicates NTH plays a crucial role in removal of oxidative base lesions in mitochondrial DNA.
Especificaciones
Especificaciones
| Antígeno | NTHL1 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Recombinant Monoclonal |
| Clon | 8P6Z4 |
| Concentración | 1.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | NTHL1 |
| N.º de referencia del gen | P78549 |
| Alias de gen | bifunctional DNA N-glycoslyase/DNA-(apurinic or apyrimidinic site) lyase; Bifunctional DNA N-glycosylase/DNA-(apurinic or apyrimidinic site) lyase; DNA glycoslyase/AP lyase; DNA glycosylase/AP lyase; Endonuclease III-like protein 1; FAP3; hNTH1; nth (endonuclease III)-like 1 (E.coli); nth endonuclease III-like 1; Nth1; NTHL 1; Nthl1; nth-like DNA glycosylase 1; OCTS 3; OCTS3; thymine glycol DNA glycosylase/AP lyase |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?