missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NTAL Monoclonal Antibody (NAP-07), PE
Descripción
This antibody reacts with an intracellular epitope of Non-T cell activation linker (NTAL), also known as LAB (linker of activated B cells), a 25 - 30 kDa transmembrane adaptor protein associating with lipid raft components in the plasma membrane of B lymphocytes, NK cells and myeloid cells.
NTAL (Non-T cell Activation Linker), also known as LAB (Linker for activation of B cells), is a 30 kDa double-palmitoylated transmembrane adaptor protein expressed by B cells, NK cells, mast cells and macrophages. It is a negative regulator of early stages of BCR-dependent B cell signaling and serves as a negative regulator also in mast cells. However, in mast cells, NTAL also contributes to some activation processes, partially overlapping with LAT function.
Especificaciones
Especificaciones
| Antígeno | NTAL |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | NAP-07 |
| Concentración | 0.1 mg/mL |
| Conjugado | PE |
| Formulación | PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 |
| génica | LAT2 |
| N.º de referencia del gen | Q9GZY6, Q9JHL0 |
| Alias de gen | AW125574; HSPC046; LAB; Lat2; LAT-2 like; Linker for activation of B-cells; linker for activation of T cells family, member 2; linker for activation of T cells, transmembrane adaptor 2; linker for activation of T-cells family member 2; Membrane-associated adapter molecule; non-T cell activation linker; non-T-cell activation linker; NTAL; WBS15; WBSCR15; WBSCR5; Williams-Beuren syndrome chromosomal region 15 protein; williams-Beuren syndrome chromosomal region 15 protein homolog; Williams-Beuren syndrome chromosomal region 5 protein; Williams-Beuren syndrome chromosome region 5 homolog; Williams-Beuren syndrome chromosome region 5-like protein; WSCR5 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?