Learn More
NSUN2 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Código de producto. 18044495 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
100 μL
20 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
18044495 20 μL 20 microlitros
18085175 100 μL 100 microlitros
2 options
Código de producto. 18044495
Marca: Proteintech CL4882085420UL
Rabbit Polyclonal Antibody
The NSUN2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NSUN2. This antibody recognizes human antigen. The NSUN2 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Especificaciones
Antígeno NSUN2
Aplicaciones Flow Cytometry (Intracellular Staining)
Concentración 1000 μg/mL
Conjugado CoraLite Plus 488
Símbolos de los genes FLJ20303, hTrm4, MISU, NSUN2, SAKI, TRM4
Especie del huésped Rabbit
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 54888
Especies diana Human
Contenido y almacenamiento Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Peso molecular del anticuerpo 150 kDa
Formulario Liquid
Isotype IgG