missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSL1 Monoclonal Antibody (OTI4D3), TrueMAB™, OriGene
Descripción
Part of the MIS12 complex which is required for normal chromosome alignment and segregation and kinetochore formation during mitosis.
Especificaciones
Especificaciones
| Antígeno | NSL1 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI4D3 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | NSL1 |
| N.º de referencia del gen | Q96IY1 |
| Alias de gen | C1orf48, DC8, MIS14 |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?