Learn More
NRF1/nuclear respiratory factor 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16800224 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16800224 150 μL 150 microlitros
16810224 20 μL 20 microlitros
2 options
Código de producto. 16800224
Marca: Proteintech 124821AP150UL
Rabbit Polyclonal Antibody
This gene encodes a protein that homodimerizes and functions as a transcription factor which activates the expression of some key metabolic genes regulating cellular growth and nuclear genes required for respiration, heme biosynthesis, and mitochondrial DNA transcription and replication. The protein has also been associated with the regulation of neurite outgrowth. Alternate transcriptional splice variants, which encode the same protein, have been characterized. Additional variants encoding different protein isoforms have been described but they have not been fully characterized. Confusion has occurred in bibliographic databases due to the shared symbol of NRF1 for this gene and for ""nuclear factor (erythroid-derived 2)-like 1"" which has an official symbol of NFE2L1.
Especificaciones
Antígeno NRF1/nuclear respiratory factor 1
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.19 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica NRF1
N.º de referencia del gen Q16656, Q62792, Q9WU00
Alias de gen ALPHA PAL, NRF 1, NRF1, NRF1/nuclear respiratory factor 1, nuclear respiratory factor 1
Símbolos de los genes Nrf1
Especie del huésped Rabbit
Inmunógeno NRF1/nuclear respiratory factor 1 Fusion Protein Ag3172
Método de purificación Antigen Affinity Chromatography
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 18181, 312195, 4899
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.