missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NRF1 Monoclonal Antibody (2G4)
GREENER_CHOICE

Código de producto. 17136733 Tienda Thermo Scientific Productos
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17136733 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17136733 Proveedor Invitrogen™ N.º de proveedor MA536224

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Jurkat whole cell, human HL-60 whole cell, human K562 whole cell, human Raji whole cell, human SH-SY5Y whole cell, human THP-1 whole cell. IHC: human rectal cancer tissue, human lung cancer tissue, human mammary cancer tissue, human testis cancer tissue, human testis cancer tissue, human testicular cancer tissue, human tonsil tissue, human gastric cancer tissue, human rectal cancer tissue, human appendicitis tissue, human appendicitis tissue, human bladder cancer tissue, human lung cancer tissue, human lung cancer tissue, human melanoma tissue, human ovarian cancer tissue, human renal carcinoma tissue, human skin cancer tissue. ICC/IF: MCF7 cell. Flow: PC-3 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

This gene encodes a protein that homodimerizes and functions as a transcription factor which activates the expression of some key metabolic genes regulating cellular growth and nuclear genes required for respiration, heme biosynthesis, and mitochondrial DNA transcription and replication. The protein has also been associated with the regulation of neurite outgrowth. Alternate transcriptional splice variants, which encode the same protein, have been characterized. Additional variants encoding different protein isoforms have been described but they have not been fully characterized. Confusion has occurred in bibliographic databases due to the shared symbol of NRF1 for this gene and for 'nuclear factor (erythroid-derived 2)-like 1' which has an official symbol of NFE2L1.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno NRF1
Aplicaciones Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Clasificación Monoclonal
Clon 2G4
Concentración 500 μg/mL
Conjugado Unconjugated
Formulación PBS with 4mg trehalose and 0.05mg sodium azide
génica NRF1
N.º de referencia del gen Q16656
Alias de gen Alpha palindromic-binding protein; alpha-pal; C87038; D6Ertd415e; EWG; LOW QUALITY PROTEIN: nuclear respiratory factor 1; Nfe2l1_retired; Not really finished protein; nrf; Nrf1; NRF-1; nrf1 protein; nrl; nuclear respiratory factor 1; nuclear respiratory factor-1; SC:BZ3C13.1; SC:bZ3C13.2; transcription factor; ube2h
Símbolos de los genes NRF1
Especie del huésped Mouse
Inmunógeno E. coli-derived human NRF1 recombinant protein (Position: D246-Q503).
Método de purificación Affinity chromatography
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 4899
Especies diana Human, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Lyophilized
Isotype IgG2a
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.