Learn More
NRAS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16857293 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
16857293 20 μL 20 microlitros
16847293 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 16857293 missing translation for 'mfr' Proteintech missing translation for 'supplierNo' 107241AP20UL
This item is not returnable. View return policy
Rabbit Polyclonal Antibody
NRAS (GTPase NRas) is a membrane protein that shuttles between the Golgi apparatus and the plasma membrane. This shuttling is regulated through palmitoylation and depalmitoylation by the ZDHHC9-GOLGA7 complex. This protein, which has intrinsic GTPase activity, is activated to a GTP-bound form by a GTPase activating protein and inactivated to a GDP-bound form by a guanine nucleotide-exchange factor. Defects in the gene encoding this protein are a cause of juvenile myelomonocytic leukemia (JMML) and Noo syndrome 6.
Specifications
Antígeno NRAS
Aplicaciones Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Flow Cytometry, Immunocytochemistry
Clasificación Polyclonal
Concentración 0.27 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica NRAS
N.º de referencia del gen P01111, P08556, Q04970
Alias de gen ALPS4, GTPase NRas, HRAS1, N ras, NRAS, NRAS1, Transforming protein N Ras
Símbolos de los genes Nras
Especie del huésped Rabbit
Inmunógeno NRAS Fusion Protein Ag1081
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 18176, 24605, 4893
Especies diana Human, Rat, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG