missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NR1I2 Mouse anti-Human, Clone:1D12G1, Abnova™
Mouse monoclonal antibody raised against recombinant human NR1I2.
Marca: Abnova MAB17263.100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This gene product belongs to the nuclear receptor superfamily, members of which are transcription factors characterized by a ligand-binding domain and a DNA-binding domain. The encoded protein is a transcriptional regulator of the cytochrome P450 gene CYP3A4, binding to the response element of the CYP3A4 promoter as a heterodimer with the 9-cis retinoic acid receptor RXR. It is activated by a range of compounds that induce CYP3A4, including dexamethasone and rifampicin. Several alternatively spliced transcripts encoding different isoforms, some of which use non-AUG (CUG) translation initiation codon, have been described for this gene. Additional transcript variants exist, however, they have not been fully characterized. [provided by RefSeq]Especificaciones
| NR1I2 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry Immunocytochemistry (1:200-1:1000) Flow Cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
|
| NR1I2 | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA, Flow Cytometry, Immunocytochemistry, Western Blot | |
| 1D12G1 | |
| nuclear receptor subfamily 1, group I, member 2 | |
| In PBS (0.05% sodium azide) | |
| BXR, ONR1, PAR, PAR1, PAR2, PARq, PRR, PXR, SAR, SXR | |
| Recombinant protein corresponding to amino acid 1-142 of human NR1I2 from E. coli. | |
| 8856 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido