missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NR1H4 Mouse anti-Human, Clone: OTI4F12, liquid, TrueMAB™
Mouse Monoclonal Antibody
Marca: Origene Technologies TA809306
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The steroid receptor superfamily acts through direct association with DNA sequences known as hormone response elements (HREs) and bind DNA as either homo- or heterodimers. The promiscuous mediator of heterodimerization, RXR, is the receptor for 9-cis retinoic acid, and dimerizes with VDR, TR, PPAR, as well as several novel receptors including LXR (also referred to as RLD-1) and FXR. FXR and LXR fall into a category of proteins termed orphan receptors because of their lack of a defined function, and in the case of LXR, the lack of a defined ligand. FXR has been shown to bind a class of lipid molecules called farnesoids. LXR/RXR heterodimers have highest affinity for DR-4 DNA elements while FXR/RXR heterodimers bind IR-1 elements. Both LXR/RXR and FXR/RXR heterodimers retain their responsiveness to 9-cis retinoic acid.Especificaciones
| NR1H4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96RI1 | |
| NR1H4 | |
| Human recombit protein fragment corresponding to amino acids 21-297 of human NR1H4 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4F12 | |
| Unconjugated | |
| NR1H4 | |
| BAR, FXR, HRR-1, HRR1, PFIC5, RIP14 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9971 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Corrección del contenido de un producto
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido