missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NQO2 Monoclonal Antibody (OTI4B2), TrueMAB™, OriGene
Description
NQO2 (EC 1.10.99.2) is a flavoprotein that catalyzes the 2-electron reduction of various quinones, redox dyes, and the vitamin K menadione. NQO2 predomitly uses dihydronicotinamide riboside (NRH) as the electron donor.
Specifications
Specifications
| Antígeno | NQO2 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI4B2 |
| Concentración | 0.9 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | NQO2 |
| N.º de referencia del gen | P16083 |
| Alias de gen | DHQV, DIA6, NMOR2, QR2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?