missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NQO2 Monoclonal Antibody (OTI4B2), TrueMAB™, OriGene
Description
NQO2 Monoclonal antibody specifically detects NQO2 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antígeno | NQO2 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI4B2 |
| Concentración | 0.9 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | NQO2 |
| N.º de referencia del gen | P16083 |
| Alias de gen | DHQV; DIA6; EC 1.10.99.2; MGC94180; NAD(P)H dehydrogenase, quinone 2; NAD(P)H menadione oxidoreductase 2, dioxin inducible; NAD(P)H menadione oxidoreductase-1, dioxin-inducible-2; NAD(P)H quinone dehydrogenase 2; NMO2; NMOR2; NQO 2; Nqo2; NRH dehydrogenase; NRH dehydrogenase [quinone] 2; NRH: quinone oxidoreductase; NRH:quinone oxidoreductase 2; N-ribosyldihydronicotinamide:quinone reductase 2; OTTHUMP00000015948; OTTHUMP00000015949; OTTHUMP00000015953; Ox 2; Ox2; QR2; Quinone; Quinone reductase 2; quinone ribosyldihydronicotinamide dehydrogenase; ribosyldihydronicotinamide; ribosyldihydronicotinamide dehydrogenase; ribosyldihydronicotinamide dehydrogenase [quinone]; ribosyldihydronicotinamide dehydrogenase quinone |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?