missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NQO-2 Antibody (1E3), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP1-74033UV
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
NQO-2 Monoclonal specifically detects NQO-2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Especificaciones
| NQO-2 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NQO2 | |
| Recombinant human NQO2 (1-231aa) purified from E. coli | |
| 100 μL | |
| Stem Cell Markers | |
| 4835 | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA | |
| 1000 | |
| Western Blot, ELISA | |
| dioxin-inducible-2, EC 1.10.99.2, NAD(P)H dehydrogenase, quinone 2, NAD(P)H menadione oxidoreductase 2, dioxin-inducible, NRH dehydrogenase [quinone] 2, NRH:quinone oxidoreductase 2, Quinone reductase 2, ribosyldihydronicotinamide dehydrogenase, ribosyldihydronicotinamide dehydrogenase [quinone] | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human NQO2 mAb, clone 1E3, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human NQO2 protein. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido