missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NQO-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-19573
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
NQO-1 Polyclonal specifically detects NQO-1 in Human samples. It is validated for Western Blot.
Especificaciones
| NQO-1 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 10% Glycerol with 0.01% Thimerosal | |
| Azoreductase, DHQU, DIA4azoreductase, diaphorase (NADH/NADPH) (cytochrome b-5 reductase), dioxin-inducible 1, DTDEC 1.6.5.2, DT-diaphorase, Menadione reductase, NAD(P)H dehydrogenase [quinone] 1, NAD(P)H dehydrogenase, quinone 1, NAD(P)H:menadione oxidoreductase 1, NAD(P)H:Quinone acceptor oxidoreductase type 1, NAD(P)H:quinone oxidoreductase 1, NAD(P)H:quinone oxireductase, NMOR1diaphorase-4, Phylloquinone reductase, QR1NMORI, Quinone reductase 1 | |
| Rabbit | |
| 31 kDa | |
| 0.1 mL | |
| Cancer, Cellular Markers, Lipid and Metabolism, Signal Transduction | |
| 1728 | |
| Human | |
| IgG |
| Western Blot | |
| 0.2mg/mL | |
| Western Blot 1:500-1:3000 | |
| P15559 | |
| NQO1 | |
| Recombinant protein encompassing a sequence within the center region of human NQO1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Species reactivity based on sequence identity: Mouse (86%), Pig (89%), Rat (85%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido