Learn More
Novus Biologicals™ Human Brain Medulla Oblongata Whole Tissue Lysate (Adult Normal)
Marca: Novus Biologicals™ NB820-59189
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The Human Brain Medulla Oblongata Whole Tissue Lysate (Adult Whole Normal) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.Especificaciones
| Immunoprecipitation, Western Blot | |
| 1 mg | |
| Total Protein - Human Adult Normal Tissue: Brain: Medulla oblongata | |
| Normal | |
| Human | |
| -80°C. Avoid freeze-thaw cycles. |
| Nervous | |
| HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors | |
| Whole | |
| Human | |
| 5 mg/mL | |
| Native |